Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Aminopropan-1-ol ≥99.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd
Danger

A0438-500G A0438-25G
TCA0438-500GEA 115.1 USD InStock
TCA0438-500G TCA0438-25G
3-Aminopropan-1-ol ≥99.0% (by GC, titration analysis)
3-Aminopropan-1-ol
CAS Number: 156-87-6
MDL Number: MFCD00008223
Molecular Formula: C3H9NO
Molecular Weight: 75.11
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 188
Flash Point (°C): 101
Freezing point (°C): 11
Specific Gravity (20/20): 0.99

Other Product Names / Synonyms: Propanolamine


Formula: C₃H₉NO
Boiling Pt: 188 °C
Melting Pt: 11 °C
Density: 0.99 (20 °C)
Flash Pt: 101 °C
MDL Number: MFCD00008223
CAS Number: 156-87-6
EINECS: 205-864-4
UN: 2735
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b6c3f5424029e4a4efd18a67ff32aad07703709&vid=7b6c3f5424029e4a4efd18a67ff32aad07703709&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty