Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzo[a]pyrene ≥95.0%, purified by sublimation

Supplier: Decon Laboratories Ltd
Danger

B0085-100MG B0085-1G
TCB0085-100MGEA 55.93 USD InStock
TCB0085-100MG TCB0085-1G
Benzo[a]pyrene ≥95.0%, purified by sublimation
Benzo[a]pyrene
CAS Number: 50-32-8
MDL Number: MFCD00003602
Molecular Formula: C20H12
Molecular Weight: 252.32
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 496
Melting point (°C): 176

Product Categories: Materials Chemistry, Nanocarbon Unit Structures (Nanocarbon Materials), Highly Purified Reagents, Bioscience, Reagents for Carcinogenic Models, Analytical Science, Estradiol, etc. (Environmental Endocrine Disruptors)
Other Product Names / Synonyms: Benzo[a]pyrene


Formula: C₂₀H₁₂
Boiling Pt: 496 °C
Melting Pt: 176 °C
Density: 1.351 g/cm³ (20 °C)
MDL Number: MFCD00003602
CAS Number: 50-32-8
EINECS: 200-028-5
UN: 2811
ADR: 6.1,III
Merck Index: 12,01134
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b5174e0a8dd97e7fb65c88d9b178daeadf5dfb7&vid=7b5174e0a8dd97e7fb65c88d9b178daeadf5dfb7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty