Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bromocyclopentane ≥98.0%

Supplier: Decon Laboratories Ltd

B1006-25G B1006-500G
TCB1006-25GEA 47 USD InStock
TCB1006-25G TCB1006-500G
Bromocyclopentane ≥98.0%
Bromocyclopentane
CAS Number: 137-43-9
MDL Number: MFCD00001359
Molecular Formula: C5H9Br
Molecular Weight: 149.03
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 138
Flash Point (°C): 35
Specific Gravity (20/20): 1.39

Other Product Names / Synonyms: Cyclopentyl Bromide


Formula: C₅H₉Br
Boiling Pt: 138 °C
Density: 1.39 (20 °C)
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00001359
CAS Number: 137-43-9
EINECS: 205-294-6
UN: 1993
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d91c1f7ec0a8b62591c814e4518a45880174940&vid=0d91c1f7ec0a8b62591c814e4518a45880174940&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty