Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloropyridine ≥98.0%

Supplier: Decon Laboratories Ltd

C0279-500G C0279-25G
TCC0279-500GEA 100 USD InStock
TCC0279-500G TCC0279-25G
2-Chloropyridine ≥98.0%
2-Chloropyridine
CAS Number: 109-09-1
MDL Number: MFCD00006228
Molecular Formula: C5H4ClN
Molecular Weight: 113.54
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 170
Flash Point (°C): 67
Freezing point (°C): -47
Specific Gravity (20/20): 1.21


Formula: C₅H₄ClN
Boiling Pt: 170 °C
Melting Pt: –46 °C
Density: 1.21 (20 °C)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00006228
CAS Number: 109-09-1
EINECS: 203-646-3
UN: 2822
ADR: 6.1,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0a8b21245391eb89ed37821cad5c40835ae57e4&vid=c0a8b21245391eb89ed37821cad5c40835ae57e4&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty