Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Difluorobenzaldehyde ≥96.0%

Supplier: Decon Laboratories Ltd
Warning

D2637-5G D2637-25G
TCD2637-005GEA 49 USD InStock
TCD2637-005G TCD2637-25G
2,5-Difluorobenzaldehyde ≥96.0%
2,5-Difluorobenzaldehyde
CAS Number: 2646-90-4
MDL Number: MFCD00010327
Molecular Formula: C7H4F2O
Molecular Weight: 142.10
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 66
Flash Point (°C): 58
Specific Gravity (20/20): 1.31


Formula: C₇H₄F₂O
Boiling Pt: 66 °C
Density: 1.31 (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00010327
CAS Number: 2646-90-4
UN: 1989
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64aa18180425633b1b32ce4680015b47704cdcf8&vid=64aa18180425633b1b32ce4680015b47704cdcf8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty