Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl ethylmalonate ≥98.0%

Supplier: Decon Laboratories Ltd

E0138-25ML E0138-500ML
TCE0138-025MLEA 21.37 USD InStock
TCE0138-025ML TCE0138-500ML
Diethyl ethylmalonate ≥98.0%
Diethyl ethylmalonate
CAS Number: 133-13-1
MDL Number: MFCD00009167
Molecular Formula: C9H16O4
Molecular Weight: 188.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 211
Flash Point (°C): 88
Specific Gravity (20/20): 1.01

Other Product Names / Synonyms: Ethylmalonic Acid Diethyl Ester


Formula: C₉H₁₆O₄
MW: 188.22 g/mol
Boiling Pt: 211 °C
Density: 1.01 (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD00009167
CAS Number: 133-13-1
EINECS: 205-093-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5e6caf6d0ba9a8f3c9c6592845b5a272b6591cf&vid=f5e6caf6d0ba9a8f3c9c6592845b5a272b6591cf&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty