F0379-25G
F0379-5G
TCF0379-25GEA
306.5
USD
InStock
TCF0379-25G
TCF0379-005G
3-(4-Fluorobenzoyl)propionic acid ≥97.0%
3-(4-Fluorobenzoyl)propionic acid
CAS Number: 366-77-8
MDL Number: MFCD00002793
Molecular Formula: C10H9FO3
Molecular Weight: 196.18
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 102