Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexamethyldisilane ≥98.0%

Supplier: Decon Laboratories Ltd

H0638-100ML H0638-10ML
TCH0638-100MLEA 202.42 USD InStock
TCH0638-100ML TCH0638-10ML
Hexamethyldisilane ≥98.0%
Hexamethyldisilane
CAS Number: 1450-14-2
MDL Number: MFCD00008258
Molecular Formula: C6H18Si2
Molecular Weight: 146.38
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 113
Flash Point (°C): -9
Freezing point (°C): 13
Specific Gravity (20/20): 0.73


Formula: C₆H₁₈Si₂
MW: 146.38 g/mol
Boiling Pt: 112
Melting Pt: 13…15 °C
Density: 0.73 (20 °C)
Flash Pt: –9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008258
CAS Number: 1450-14-2
EINECS: 215-911-0
UN: 1993
ADR: 3 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44f58d1121bad72aef71c0da180e07dcfff3e856&vid=44f58d1121bad72aef71c0da180e07dcfff3e856&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty