2-Octene ≥98.0%

Supplier: Decon Laboratories Ltd

O0042-25ML O0042-500ML
TCO0042-025MLEA 73 USD
TCO0042-025ML TCO0042-500ML
2-Octene ≥98.0%
2-Octene

CAS Number: 111-67-1
MDL Number: MFCD00009532
Molecular Formula: C8H16
Molecular Weight: 112.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 125
Flash Point (°C): 21
Specific Gravity (20/20): 0.72


Other Product Names / Synonyms: 2-Octylene

Formula: C₈H₁₆
MW: 112.22 g/mol
Boiling Pt: 125 °C
Melting Pt: -106.8
Density: 0.72 (20 °C)
Flash Pt: 21 °C
MDL Number: MFCD00009532
CAS Number: 111-67-1
EINECS: 203-894-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d27e5737540d6642ac49357d7d30b6dcf1063dc&vid=6d27e5737540d6642ac49357d7d30b6dcf1063dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR