T0026-500G
T0026-25G
T0026-100G
TCT0026-500GEA
315.3
USD
InStock
TCT0026-500G
TCT0026-025G
TCT0026-100G
D-(-)-Tartaric acid ≥99.0%
D-(-)-Tartaric acid
CAS Number: 147-71-7
MDL Number: MFCD00004238
Molecular Formula: C4H6O6
Molecular Weight: 150.09
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Color: White
Specific rotation [a]20/D: -13 deg (C=20, H2O)