Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Toluenethiol ≥97.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd
Warning

T0290-500G T0290-25G
TCT0290-500GEA 363 USD InStock
TCT0290-500G TCT0290-025G
4-Toluenethiol ≥97.0% (by GC, titration analysis)
4-Toluenethiol
CAS Number: 106-45-6
MDL Number: MFCD00004851
Molecular Formula: C7H8S
Molecular Weight: 124.20
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 196
Melting point (°C): 43
Flash Point (°C): 68

Other Product Names / Synonyms: 4-Mercaptotoluene, p-Thiocresol, p-Tolyl Mercaptan, 4-Methylbenzenethiol


Formula: C₇H₈S
Boiling Pt: 196 °C
Melting Pt: 43 °C
Density: 1.022 g/cm³ (51 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00004851
CAS Number: 106-45-6
EINECS: 203-399-1
UN: 2811
ADR: 6.1,III
Merck Index: 12,09461
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=917f0754391973827e482aee051592aced65dc9a&vid=917f0754391973827e482aee051592aced65dc9a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty