Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tributylamine ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd
Danger

T0357-500ML T0357-25ML
TCT0357-500MLEA 32.4 USD InStock
TCT0357-500ML TCT0357-25ML
Tributylamine ≥98.0% (by GC, titration analysis)
Tributylamine
CAS Number: 102-82-9
MDL Number: MFCD00009431
Molecular Formula: C12H27N
Molecular Weight: 185.36
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 217
Melting point (°C): -70
Flash Point (°C): 74
Specific Gravity (20/20): 0.78


Formula: C₁₂H₂₇N
Boiling Pt: 217 °C
Melting Pt: –70 °C
Density: 0.78 (20 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00009431
CAS Number: 102-82-9
EINECS: 203-058-7
UN: 2542
ADR: 6.1,II
Merck Index: 13,09691
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=241763504005ba63b2472e70d918896745887279&vid=241763504005ba63b2472e70d918896745887279&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty