Magnetic 3D Cell Culturing
3D Cell Culture
Magnetic 3D Cell Culturing 3D in a fast and easy 2D workflow 
The core technology is the magnetization of cells with NanoShuttle™-PL. The cells can be aggregated with magnetic forces, either by levitation or printing, to form structurally and biologically representative 3D models in vitro.
  • The advantages of magnetic cell culture include
  • Mimicking native tissue environment
  • Rapid 3D model formation within hours
  • No specialized equipment, media, or artificial substrate
  • Easy to handle / no sample loss
  • Allows co-culture
CELLSTAR® Cell Culture Vessels with Cell-Repellent Surface
In contrast to standard tissue culture surfaces which are optimized to enhance conditions for cell attachment, the cell-repellent surface has been developed to effectively prevent cell attachment. CELLSTAR® cell culture vessels with a cell-repellent surface reliably prevent cell attachment in suspension cultures of semi adherent and adherent cell lines where standard hydrophobic surfaces generally used for suspension culture are insufficient.
Magnetic 3D Cell Culture Products
VWR PN GBO PN Product Descriptions
76324-742 657840 6-Well Bio-Assembler Kit
76324-760 662840 24-Well Bio-Assembler™ Kit
76324-014 655840 96-Well Bioprinting Kit, Clear Plates
76324-016 655841 96-Well Bioprinting Kit, Black µClear Plates
76324-018 655846 96-Well BiO Assay™ Kit
76324-020 655849 96-Well Bio Assay Kit™ & Imaging System
76324-768 781840 384-Well Bioprinting Kit, Clear Plates
76324-770 781841 384-Well Bioprinting Kit, Black µClear Plates
76324-772 781846 384-Well BiO Assay™ Kit
76324-282 781849 384-Well BiO Assay™ Kit & Imaging System
76337-248 781850 384-Well Ring Drive
76337-250 789830 1536 well Spheroid Drive
76337-252 789837 1536 WELL HOLDING DRIVE
76324-744 657841 NanoShuttle™-PL Refill, 600 µL Vial (1)
76324-746 657843 NanoShuttle™-PL Refill 3 Pack, 600 µL Vials (3)
76324-748 657846 NanoShuttle™-PL Refill 6 Pack, 600 µL Vials (6)
76324-750 657847 NanoShuttle™-PL Refill 6 Pack with FREE iPod™ (Only with purchase of BIO Assay™ 655849 or 781849)
76324-754 657852 NanoShuttle™-PL Refill 12 pack, 600 µl Vials
76324-752 657850 MagPen™ 3 Pack
76324-024 657860 Imaging Kit
CELLSTAR® Cell Culture Vessels with Cell-Repellent Surface
VWR PN GBO PN Product Descriptions
10014-312 627979 CELLSTAR 35mm dish, cell-repellent surface, clear, sterile, with lid
10014-314 628979 CELLSTAR® Cell-Repellent Surface, 60x15mm, sterile, with lid
30618-024 664970 Cellstar 100mm dish, cell-repellent surface, clear, sterile, with lid
75838-906 690980 CELL CULTURE FLASK, 50 ML, 25 CM², PS, CLEAR,CELLSTAR®, CELL-REPELLENT SURFACE, WHITE STANDARD SCREW CAP, STERILE, 10 PCS./BAG
10836-539 690985 T25 CELLSTAR® Cell-Repellent Surface, sterile, filter cap
75838-908 658980 CELL CULTURE FLASK, 250 ML, 75 CM², PS,CLEAR, CELLSTAR®, CELL-REPELLENT SURFACE, WHITE STANDARD SCREW CAP, STERILE, 5 PCS./BAG
10836-541 658985 T75 CELLSTAR® Cell-Repellent Surface, sterile, filter cap
75845-114 660980 CELL CULTURE FLASK, 550 ML 175 CM², PS, Clear, CELLSTAR®, Cell Repellent Surface, White Standard Screw Cap
75800-006 660985 CELL CULTURE FLASK, 550 ML, 175 CM², PS, Clear, CELLSTAR®, Cell Repellent Surface, White Filter Screw Cap, Sterile
75838-910 661980 CELL CULTURE FLASK, 650 ML, 175 CM², PS,HIGH FLASK DESIGN, CLEAR, CELLSTAR®, CELL-REPELLENT SURFACE, WHITE STANDARD SCREW CAP,STERILE, 4 PCS./BAG
30618-022 657970 Cellstar 6 well plate, cell-repellent surface, clear, sterile, with lid
10014-320 662970 24 Well CELLSTAR® Cell-Repellent Surface, sterile, with lid
10014-322 677970 48 Well CELLSTAR® Cell-Repellent Surface, sterile, with lid
30618-028 655970 96W Cell-Repellent Plate, PS, Sterile, Flat Bottom, Chimney Style, Clear, w/Lid
10014-318 655976 96W Cell-Repellent Plate, PS, Sterile, Flat Bottom, Black µClear, w/Lid
10859-852 655976-SIN 96W Cell-Repellent Plate, PS, Sterile, Flat Bottom, Black µClear, w/Lid
30618-026 650970 96W Cell-Repellent Plate, PS, Sterile, Round (U) Bottom, Clear, w/Lid
76199-200 650979 96W Cell-Repellent Plate, PS, Sterile, Round (U) Bottom, Clear, w/Lid
10014-316 651970 96W Cell Repellent Plate, PS, Sterile, Conical (V) Bottom, Clear, w/Lid
10014-324 781970 384W Cell-Repellent Plate, PS, Sterile, Flat Bottom, Clear, w/Lid
76371-716 781974 384W Cell-Repellent Plate, PS, Sterile, Flat Bottom, White µClear, w/Lid
10014-326 781976 384W Cell-Repellent Plate, PS, Sterile, Flat Bottom, Black µClear, w/Lid
10859-850 781976-SIN 384W Cell-Repellent Plate, PS, Sterile, Flat Bottom, Black µClear, w/Lid
76199-198 787979 384 Well Cell-Repellent plate, PS, Sterile, U Bottom, clear w/lid