Product Differentiation Table
|
FBS Products |
FBS Alternatives |
Ultimate Grade FBS |
Premium Grade FBS |
USDA Approved Origin FBS |
Ultra Low IgG FBS |
FB Essence |
Bovine Calf Serum, Iron Supplemented |
Country of Origin |
100% US Origin |
X |
X |
|
X |
X |
X |
USDA Approved Origin (Non-US) |
|
|
X |
|
|
|
Product Features |
Single-Use Filtration Technology |
X |
X |
X |
X |
X |
|
Cold Temperature Manufactured |
X |
X |
X |
|
X |
|
Class 100 Cleanroom Manufactured |
X |
X |
X |
|
X |
|
Triple 0.1μm Sterile Filtered |
X |
X |
X |
X |
X |
|
Whole-lot Homogenous Technology |
X |
X |
X |
X |
X |
X |
Independently Tested by Third-Party Laboratories |
X |
X |
X |
X |
X |
X |
Specifications |
ISIA† Traceability Certified Sera |
X |
X |
X |
X |
X |
X |
Endotoxin** |
≤ 10 |
≤ 20 |
≤ 30 |
≤ 20 |
≤ 20 |
≤ 20 |
Hemoglobin** |
≤ 25 |
≤ 25 |
≤ 30 |
≤ 25 |
≤ 25 |
≤ 30 |
9CFR Virus Testing, Sterility, Mycoplasma |
X |
X |
X |
X |
X |
X |
Biochemical Assay |
X |
X |
X |
X |
X |
X |
Electrophoretic Profile |
X |
X |
X |
X |
X |
X |
Additional Virus Testing by qPCR Technology |
X |
|
|
|
|
|
† International Serum Industry Association (serumindustry.org)
** The care and attention devoted to the manufacture of our sera generates final test results that are substantially lower than set specifications. See lot-specific Certificates of Analysis for actual test results.