Harleco & Midas - Made for Each Other
Clinical Microscopy
Microscopy products to support your success.
Related Information:
HARLECO® Dyes & Stains
  • Highest Quality Stain Solutions - IVD certified, also certified by the Biological Stain Commission
  • Ready to use staining solutions for blood and bone marrow samples with intense and reproducible results
  • Very clean results in less than 1 minute with Hemacolor®
The Midas® III-Plus Automated Stainer
  • The six station unit holds 20 or 30 slides
  • Up to 10 programs - each program can have up to 29 steps
  • Save up to 50% on stain consumption vs. flood or spray techniques