Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD44 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: F10-44-2]

Supplier: Southern Biotechnology
9400-09 9400-09S
100245-974EA 347.85 USD InStock
100245-974 103260-154
Anti-CD44 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: F10-44-2]
Antibodies
CD44, also known as phagocytic glycoprotein-1 (Pgp-1), exists as a large number of different isoforms resulting from alternative RNA splicing. The major isoform expressed on lymphocytes, myeloid cells, and erythrocytes is a glycosylated type I transmembrane protein. Other variable isoforms containing glycosaminoglycans have molecular weights ranging from 110 to 250 kDa and are widely expressed on hematopoietic and non-hematopoietic cells. CD44 contributes to the adhesion of leukocytes to endothelial cells, stromal cells, and the extracellular matrix. It also plays a functional role in cell migration, lymphocyte homing, and adhesion during hematopoiesis and lymphocyte activation.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: F10-44-2
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Ma's Night Monkey

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty