Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD14 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 61D3]

Supplier: Southern Biotechnology
9561-09 9561-09S
100246-170EA 315.15 USD InStock
100246-170 103265-322
Anti-CD14 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 61D3]
Antibodies
CD14 is a 55 kDa glycosphosphatidylinositol (GPI)-anchored single chain glycoprotein. It is expressed on monocytes, macrophages, dendritic cells, and Langerhans cells. It is also weakly expressed on neutrophils. CD14 appears to be involved in clearance of gram-negative bacteria via its high affinity binding to LPS-LPB complexes.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Cynomolgus, Canine, Hooded Seal

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty