Anti-ZAP-70 Mouse Monoclonal Antibody (Biotin) [clone: SBZAP]

Supplier: Southern Biotechnology
10650-08S 10650-08L
100242-628EA 129.46 USD
100242-628 100242-626
Anti-ZAP-70 Mouse Monoclonal Antibody (Biotin) [clone: SBZAP]
Antibodies
ZAP-70 is a protein tyrosine kinase (PTK) with the primary role of catalyzing the phosphorylation of tyrosine residues in cellular proteins, and thus, contributes significantly to the initiation of cell signaling. ZAP-70 is normally expressed on T cell and NK cells.

Type: Primary
Antigen: ZAP-70
Clonality: Monoclonal
Clone: SBZAP
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR