2,3-Dimethoxyaniline 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A839404-1G A839404-250MG A839404-5G A839404-25G
77171-596EA 24 USD
77171-596 77171-598 77171-600 77684-801
2,3-Dimethoxyaniline 98%, Ambeed.Inc
2,3-Dimethoxyaniline

Precaución: For research use only.

Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
MDL Number: MFCD00180769
CAS Number: 6299-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR