VWR, Part of Avantor
Carousel

Supporting Research of COVID-19
Easy access to products and protocols for research use only in the identification of 2019-nCoV based on Centers for Disease Control and Prevention (CDC) recommendations

Managing Supplies and Inventory for Over 160 Years
Your inventory should benefit your scientific initiative, not delay it – and if your supply management is not at its most efficient, your staff could be spending valuable time just trying to get the products they need.

Moving Forward. Together.
So much has changed during this unprecedented time, except your ability to count on Avantor. We continue to set science in motion, and know safety is paramount for you and your colleagues. Stay safe with this 3-ply level 1 face mask with aluminum nose strip to conform to any size or shape face.

Improve downstream chromatography processes
New sizes available! J.T.Baker® BAKERBOND™ PROchievA™ recombinant protein A affinity chromatography resin helps increase product purity in mAbs manufacturing.
Features
-
TSB Media Bags for Aseptic Process Simulation
IV style bags for dispensing 700ml fill tryptic soy broth from a needle or spike port. Terminally autoclaved for high sterility assurance.
-
Avantor TopMixer Single-Use Open Top Mixing Systems
Modular Mixing System with top mounted mixer for use with drums featuring integrated formulation programming for your biopharma processes.
-
Avantor ACE Excel Oligo U-HPLC Columns Promotion
Buy 1 column from the list and get 1 case of qualifying VWR Chemicals BDH Biosyntheseis Reagents - free!
-
JFK UltraSafety 3-Ply Level 1 Face Masks
FaceMasks with aluminum nose strip conform to any size or shape face and provide fluid resistance up to 120mm Hg.
-
Extran® – reliable all-purpose detergents
Residue-free cleaning in laboratories & production. Extran® from MilliporeSigma Detergents for manual cleaning (MA) or machine cleaning in laboratory (AP).
-
Wells Lamont Glove Liners – protect your hands and manage disposable shortages
Reusable nylon liners in full or half finger lengths can help stretch out your supply of disposable gloves.
-
Easy-to-seal Quick Release VWR® Irradiated Tape
With low adhesive residue on plastics and other smooth surfaces in the cleanroom, this tape is ideal for plate sealing and surface marking.
-
Brady i5100 Industrial Label Printer with Product and Wire ID Software Suite
The Brady i5100 Label Printer provides the fundamental printing capabilities for the high-mix and high-volume work you need to get done every day.
Selector guides & more... View all selector guides
-
Chemical structure search
Accelerate your discovery by reducing your search time and streamlining procurement process complexities...
-
IgGy – VWR’s antibody selector
Find your antibodies using IgGy, the Antibody Selector! Quickly find the specific antibody you need from VWR's broad portfolio...
-
Pipet tip selector
VWR's Pipet Tip Selector Guide will lead you quickly to the tip we recommend. You can search by pipettor brand or tip attribute, such as volume range, sterility, etc...
New products
-
Ansell BioClean™ Clearview Autoclavable Panoramic Goggles
Super-soft frame Indirect ventilation system, Toughened polycarbonate, anti-scratch & optically correct lens, Ideal for wearing over eye-glasses, Non-linting latex-free head band, Anti-fog performance remaining for up to 25 cycles with no degradation (under laboratory conditions)
-
Lico Spectral Colorimeters: Pharmacopeial Color Analysis of Transparent Liquids
Hach Lico Colorimeters eliminate visual analysis error, simplify color measurement, and ensure consistency in color from plant to lab to customer.
-
Performance, Reliability and Energy Efficiency
The PHCbi brand ultra-low temperature freezer product line offers a versatile selection of upright and chest models designed for a wide range of today's critical applications.
-
Avantor J.T.Baker® BAKERBOND® PROchievA™ protein A chromatography resins
High-performance protein A chromatography resins for mAbs production processes.
Literature Search all documents
-
FOCUS: Industrial
Showcases solutions critical to production in industries such as cannabis, forensics, and cosmetics
-
FOCUS: Equipment & Instruments
Highlights equipment, instrumentation, consumables and technical services for educated buying choices
-
FOCUS: Production
Features products and technical articles to make it easier to work in controlled environments
-
FOCUS: Safety
Provides the broadest range of safety supplies to keep you safe and productive across all applications