Wet/Tank Blotting System, G-Biosciences
Supplier: G-Biosciences
BT301
10001-838EA 966.26 USD InStock
10001-838
Wet/Tank Blotting System, G-Biosciences
Blotting Apparatus
In high quality acrylic with platinum electrodes, the Wet/Tank Blotting System provides high quality and rapid blotting (electro-transfer) of mini handcast and precast gels
A seamless, injection-molded transparent buffer tank prevents leakage and breakage. Suitable for rapid (one hour) transfers or overnight low-intensity transfers, the color coded cassettes and electrodes ensure easy and correct orientation and error-free set-up.
Latches on the gel holder cassette allow for easy handling, simplified insertion and removal of the gel holder cassette from supporting body for transfer (More Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty