Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ruthenium 5% on carbon

Danger

44-4050-5G 44-4050-25G
100200-276EA 65.11 USD InStock
100200-276 100196-880
Ruthenium 5% on carbon
Ruthenium


Formula: Ru
MW: 101.07
MDL Number: MFCD00011207
CAS Number: 7440-18-8
UN: 3178
ADR: 4.1 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty