Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(4-chlorophenyl)phosphine ≥99%

Warning

15-6050-1G 15-6050-5G
100204-524EA 130.22 USD InStock
100204-524 100206-092
Tris(4-chlorophenyl)phosphine ≥99%
Tris(4-chlorophenyl)phosphine
Phosphine


Formula: C₁₈H₁₂Cl₃P
MW: 365.63 g/mol
Boiling Pt: 431
Melting Pt: 103
Density: 1.449 g/cm³ (20 °C)
MDL Number: MFCD00013639
CAS Number: 1159-54-2
EINECS: 214-596-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty