Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium bromide, crystal disk for spectrophotometry, unpolished

Warning

0002N-88 0002N-396 0002N-1017 0002N-90
100215-408EA 36.25 USD InStock
100215-408 100216-120 100213-480 100215-688
Potassium bromide, crystal disk for spectrophotometry, unpolished
Potassium bromide
Unpolished crystal optics are also known as blanks. These windows are finely ground with flat parallel surfaces. As sold they are opaque. They can be water polished quickly to achieve 90% transmission. The transmission of water polished KBr, KCI and NaCl


Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435
Melting Pt: 730
Density: 2.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty