Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chlorogenic acid ≥98%, crystals

Supplier: Deelux Labortechnik GmbH

C0021GM
100268-052EA 125 USD InStock
100268-052 100268-050
Chlorogenic acid ≥98%, crystals
Chlorogenic acid
  • White crystals
  • Store in a cool, dry place


Formula: C₁₆H₁₈O₉
MW: 354.31 g/mol
Melting Pt: 208…209 °C
MDL Number: MFCD00003862
CAS Number: 327-97-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c79fd7768f68c735f0514d6459926ec541b3cabf&vid=c79fd7768f68c735f0514d6459926ec541b3cabf&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty