Dust-Off® Microfiber Cleaning Cloth, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71716-01
100493-432EA 12.68 USD InStock
100493-432 100489-354
Dust-Off® Microfiber Cleaning Cloth, Electron Microscopy Sciences
Wipes Reusable Cleaning Cloths Cloths for General Applications
For superior cleaning of delicate surfaces.
  • Environmentally responsible - lifts away oily film and dirt with no chemicals
  • Will not streak or smear surfaces
  • Non-abrasive and 100% lint-free
  • Reusable - machine or hand washable - long lasting
  • Size: 7½×7½" (19×19 cm)
Ideal for camera, lenses/filters, microscopes, computer, eyeglasses, plastic, metal, or any smooth surface.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty