Peca Techni-Valve™, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70849-10
100502-170EA 51.4 USD InStock
100502-170
Peca Techni-Valve™, Electron Microscopy Sciences
Valves Injection Valves
Peca Techni-Valve™ for use with Dust-Pro™ Refill cans.
  • Smooth bell housing and no mounting clip is required
  • Patented bell housing fits the contour of your hand
It is made from solid brass chrome plated is professionally designed with a long nozzle (90 mm (31/2")). The valve can be removed at any time without the loss of pressure.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty