Fluoromount-G™ Slide Mounting Medium, Electron Microscopy Science
Supplier: EPREDIA
17984-25
100502-406EA 112.25 USD InStock
100502-406
Fluoromount-G™ Slide Mounting Medium, Electron Microscopy Science
Stains Microscopy and staining reagents
A special mounting media for immunofluorescent staining procedures. It is a water soluble, non-fluorescing medium which is well suited for use when the staining procedure has an aqueous final step. It contains 10% polyvinyl alcohol in phosphate buffered glycerol with 0.1% sodium azide as a preservative.
A special mounting media for immunofluorescent staining procedures. It is a water soluble, non-fluorescing medium which is well suited for use when the staining procedure has an aqueous final step. It contains 10% polyvinyl alcohol in phosphate buffered glycerol with 0.1% sodium azide as a preservative.
In light microscopy, tissues were stained with conventional histological stains (e.g.,hematoxylin-eosin) and embedded in Micro-Bed: the results were comparable with those provided on paraffin embedded tissues. Further, polychrome silver-based and cytochemical stains (e.g.,PAS) were also feasible. In transmission electron microscopy, images as good as those obtained with similar acrylic resins, (e.g., Lowicryl K4M), were obtained.
Certificaciones: Certificates of analysis are available from Electron Microscopy Sciences. Please contact us or visit our website to search by lot or product number.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty