Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ruthenium(IV) oxide 0,5% aqueous solution Stabilised

Supplier: EPREDIA
Warning

20700-05
100504-374EA 651.88 USD InStock
100504-374
Ruthenium(IV) oxide 0,5% aqueous solution Stabilised
Ruthenium(IV) oxide
Ruthenium tetroxide is very similar to osmium tetroxide, and is used as a fixation/staining agent for electron microscopy.

Ruthenium tetroxide offers excellent staining of saturated and unsaturated polymer materials as well as stabilizing thin sections under the bombardment of the electron beam. Penetration of ruthenium tetroxide into the tissue is poor.

Reference: Caughey, R.C. and Miller, M.A., Proceed. of 44th Ann. Mtg. EMSA (1986), page 256, Hayat, M.A., Basic Technique for Electron Microscopy, Academic Press, NY, 1986, p 22. and Trent, J.S., Macromolecules, 17, 2930 (1984); 16, 539 (1983).


Formula: O₂Ru
MW: 133.07 g/mol
Melting Pt: 1200
Density: 1 g/cm³ (8.345 lbs/gal.)
MDL Number: MFCD00074857
CAS Number: 12036-10-1
UN: 1479
ADR: 5.1 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f4d2a220d020d6976a1906cff3e55871b52eaf9&vid=5f4d2a220d020d6976a1906cff3e55871b52eaf9&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty