Pesticide Residue Cleanup SPE Cartridges, Restek
Supplier: Restek
26193 26194
10059-518PK 399.93 USD InStock
10059-518 10059-520 10015-306
Pesticide Residue Cleanup SPE Cartridges, Restek
Chromatography Columns
Pesticide residue cleanup SPE cartridges are an efficient tool for remove matrix interferences using multiple adsorbent beds per cartridge.
  • Convenient, multiple-adsorbent beds in a single cartridge
  • For use in multiresidue pesticide analysis to remove matrix interferences
  • Excellent for cleanup of dietary supplement extracts
These cartridges deliver superior cleansing of dietary supplement extracts.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty