Amersham™ Hybond™ P Western Blotting Membrane, 0.45 PVDF, Cytiva
Supplier: GE Healthcare - Life Sciences
10600023 10600029 10600038 10600069 10600086 10600087 10600100 10600121
10061-492PK 699.01 USD InStock
10061-492 10061-494 10061-496 10061-498 10061-500 10061-502 10061-504 10061-508
Amersham™ Hybond™ P Western Blotting Membrane, 0.45 PVDF, Cytiva
Transfer Membranes
Amersham Hybond P 0.45 PVDF is a 0.45 μm pore size hydrophobic membrane, with high physical strength, highly suitable for stripping and reprobing. Membranes provide excellent performance in fluorescent and chemiluminescent applications.
  • Protein binding capacity over over 200 μg/cm² for easy signal detection
  • High protein retention even after harsh washing steps
  • High protein capture during transfer minimizes sample loss
  • Available in popular precut sizes for your application
For convenience, Western blotting sandwiches (10 sandwiches/package) areMore Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty