HF-Resistant TFE Tracey Spray Chamber for Optima 4300/5300/7300 V, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N0777497
10139-486EA 4790.16 USD InStock
10139-486
HF-Resistant TFE Tracey Spray Chamber for Optima 4300/5300/7300 V, PerkinElmer
Atomizers
This TFE Tracey™ spray chamber is used with Optima™ 4300 V, 5300 V, and 7300 V ICP-OES series instruments.
The O-ring-free Tracey spray chamber, made of tetrafluoroethylene (TFE), is designed to be hydrofluoric acid-resistant. It does not have an internal baffle feature. Its precision is considered good and its purity is considered good. They can be teamed with the PolyCon HF-resistant nebulizers to form the ideal chemically inert sample introduction system for ICP-OES.
Made from totally inert PTFE - impervious to organic solvents and strong acids, even HF Established Tracey cyclonicMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty