PTFE FAST SC Autosampler Probe, 0.8 mm I.D, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N0777523
10139-490EA 2508.41 USD InStock
10139-490
PTFE FAST SC Autosampler Probe, 0.8 mm I.D, PerkinElmer
Autosamplers
FAST probes offer high chemical resistance for applications involving strong acids, alkalis, and organics.
A contoured design reduces carryover, while an all PFA wetted sample flow path reduces contamination. SC Autosampler Probe, Carbon fiber support, 0.8mm I.D. (blue marker), long probe, long capillary, connects to port #5 (¼-28 fitting), high-flow.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty