BioFX® AP Yellow One Component Microwell Substrate (pNPP) with Stabilizer Pellets, Surmodics
Supplier: Apollo Herkenrath
PNPS-1000-01 STAB-0050-01 STAB-0020-01 STAB-0100-01 PNPS-0100-01
101397-054EA 350 USD InStock
101397-054 101397-086 101397-084 101397-088 101397-052
BioFX® AP Yellow One Component Microwell Substrate (pNPP) with Stabilizer Pellets, Surmodics
Assays ELISAs
Substrate:
The formulation consists of p-nitrophenylphosphate (p-NPP) solution with stabilizing pellets to for long term stability. The product is stable for 3 years from the date of manufacture.
  • long term stability
  • 3 year shelf life
A one-component PNPP substrate that provides a soluble yellow product upon reaction with alkaline phosphatase in ELISA applications.
Pellets:
Stabilizing Pellets for PNPS (pNPP) are chemically treated pellets for use in stabilizing phenyl phosphates.
• 5 year shelf life
Certificaciones: ISO 13485:2016 & 9001:2015 Certified
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty