Quartz Oxygen Pyrolysis Tube, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N2411235
10140-384EA 484.15 USD InStock
10140-384
Quartz Oxygen Pyrolysis Tube, PerkinElmer
Analyzers Elemental Analysers
This quartz oxygen pyrolysis tube is used with the PerkinElmer 2400 Series II Analyzer.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty