Laboratory Cold Rooms, Precision Controlled Environments, Darwin Chambers Company
Supplier: Darwin Chambers Company
VWLABG1NN08X08 VWLABG1NN10X12 VWLABG1NN10X20
10153-246EA 41795.98 USD InStock
10153-246 10153-248 10153-250
Laboratory Cold Rooms, Precision Controlled Environments, Darwin Chambers Company
Prefabricated Rooms
Darwin Laboratory Cold Rooms feature reliable, efficient environmental controls for a variety of applications
  • All rooms are constructed using 10.2 cm (4") insulated panels, R Value 31+, with white embossed aluminum interior surface finishes and white embossed galvanized steel exterior finishes
  • All include a 5.08 cm (2") insulated smooth aluminum or stainless steel floor, capable of accommodating up to 317.5 kg (700 lbs.) of evenly distributed weight
  • These rooms are modular, featuring "tongue-in-groove" construction
  • All major components are pre-wired, pre-
More Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty