Staticmaster® I-Beam Stand Ionizer and Ionizer Kit, NRD
Supplier: NRD Staticmaster
6105
10183-830EA 2137.28 USD InStock
10183-830 10183-832
Staticmaster® I-Beam Stand Ionizer and Ionizer Kit, NRD
Ionizers
The Staticmaster® 6105 I-Beam Stand Ionizer Kit is a "shockless" electrical static eliminator ideally suited for removing static charge from fine powders, filter media, and irregularly shaped items prior to being placed in the weigh chamber
  • "Shockless" electrical static eliminator
  • Useful for filter media, powders, and irregularly shaped objects
  • Removes static charges outside of weigh chamber
The I-Beam Stand Ionizer is a vertically mounted bar ionizer that is utilized outside of the weigh chamber. The Kit comes complete with an I-Beam Stand IonizerMore Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty