CONTINU-FLO Solution Set, Non Vented, Baxter IV Solutions
Supplier: Baxter
2C6571
10205-170CS 1450.98 USD InStock
10205-170
CONTINU-FLO Solution Set, Non Vented, Baxter IV Solutions
Infusions
CONTINU-FLO Solution Set with Non-Vented Spike, 0.2 Micron Filter, 2 INTERLINK Injection Sites, Backcheck Valve and Male Luer Lock Adapter Retractable Collar.
  • Contains DEHP
  • Fluid path is sterile, non pyrogenic
Approximately 10 drops/mL. Approximate Length 105" (2.7 m).
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty