Rectangular Stain Dish, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70314-01
102096-606EA 14.35 USD InStock
102096-606
Rectangular Stain Dish, Electron Microscopy Sciences
Jars Staining Jars
Heavy, clear, soda lime glass stain dish, accept glass slide measure 75 x 25 mm or 3 x 1”.
This dish comes with glass cover, holds 20 slides back-to-back horizontally.
Inside dimensions: 3 x 2 1/4x 1 1/4”.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty