High Speed Multi-Plate Shaker Vortex Mixer, MPS-1, Grant Instruments
Supplier: GRANT USA INC.
MPS-1K
10218-582EA 1626.4 USD InStock
10218-582 10218-764
High Speed Multi-Plate Shaker Vortex Mixer, MPS-1, Grant Instruments
Shakers and Mixers Microplate Shakers
Multi plate shaker and vortex can be used for tubes up to 50ml
The four platforms are: P-02-05 (24 x 0.5ml and 48 x 0.2ml); P-02-96 (96 x 0.2 ml, 8 x 0.2ml strips, 1 x PCR plate), P-05-32 (32 x 0.5ml), P-2-24 (24 x 1.5ml/2ml). Deep well plates and skirted PCR plates can be added directly to the unit. Volumes of up to 50ml can be vortexed.
The four platforms allow for semi/unskirted plates and mircotubes 0.2ml to 2ml to be mixed at up to 3200rpm. The unit also accommodates skirted PCR plates as well as deep well plates.
Certificaciones: UL Listed. CE compliant.
Información paraMore Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty