Dynamic Temperature Control Systems, Huber
Supplier: Huber
1003.0031.01 1038.0006.01 1028.0010.01 1084.0001.01 1028.0011.01 1041.0002.01 1041.0011.01 1041.0010.01 1041.0005.01 1079.0004.01 1041.0004.01 1041.0008.01 1041.0007.01 1000.0041.01 1000.0043.01 1000.0042.01 1066.0004.01 1066.0003.01
10220-788EA 26228.58 USD InStock
10220-788 10221-104 10221-106 10221-108 10221-110 10221-112 10221-114 10221-116 10221-118 10221-120 10221-728 10221-730 10221-732 10221-734 10221-736 10221-738 10221-740 10221-742 10221-744
Dynamic Temperature Control Systems, Huber
Circulating Thermostats Closed Circulator Systems Temperature Control Systems
The Unistat range inspires with unique thermodynamic properties and a range of functions to meet the highest demands
The new Grande Fleur expands the product offering in the dynamic temperature control range. The Grande Fleur together with the Petite Fleur are two small Tangos and are the entry level into the world of Unistat technology. The compact dimensions and unique thermodynamics of the Grande Fleur mean it is ideally suited for high precision temperature control of research reactors. The Grande Fleur offers all of the great performance and features of the Unistat series, achievingMore Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty