StabilZyme® Protein-Free AP Stabilizer, Surmodics
Supplier: Apollo Herkenrath
SAPF-0050 SAPF-1000
102867-012EA 43.08 USD InStock
102867-012 102867-014
StabilZyme® Protein-Free AP Stabilizer, Surmodics
Protein Linking and Labeling Systems
StabilZyme® AP and StabilZyme® Protein-Free AP stabilizers provide assay developers a complete set of tools to stabilize different AP conjugates for long term, in-solution stability.
  • Protein-free formulation
  • Extended shelf life of kits
StabilZyme® Protein-Free AP Stabilizer eliminates interference and cross-reactivity caused by proteins, delivering optimal performance for even the most sensitive immunoassays.
Certificaciones: ISO 13485:2016 &9001:2015 Certified
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty