RESISTIN ELISA, Eagle Biosciences, Inc.
Supplier: Anton Paar Hungary Kft.
E50
102994-314EA 876.15 USD InStock
102994-314
RESISTIN ELISA, Eagle Biosciences, Inc.
Assays ELISAs
Resistin ELISA measures the amount of human resistin in serum and plasma
  • Dynamic Range: 20 - 1000 pg/mL
  • Sensitivity: 12 pg/ml
  • Incubation: 4 hours
Resistin, a cysteine-rich protein of 11.3 kDa (1), was firstly found in mice (2) and constitutes together with RELMα, RELMβ and RELMγ the protein family of resistin-like molecules (RELM).  In humans, resistin and RELMβ (1) but no other proteins of the RELM family were found. The human form of resistin shows a homology of 53% to the murine protein (4). It has 11 cysteine-residues, is synthesized as a propetide of 108 amino acids and secreted as a dimer, build by a disulfide bridge of cysteine residues (22). Beside this intermolecular disulfide bridge, 5 additional intramolecular ones exist (5,6).
For research use only.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty