Transfer Membranes, Advansta
Supplier: ADVANSTA INC MS
L-08001-010 L-08012-010 L-08014-010 L-08004-010 L-08011-010 L-08013-010 L-08006-001 L-08002-010 L-08003-010
103254-726EA 111.92 USD InStock
103254-726 103254-728 103254-730 103254-732 103254-734 103254-736 103254-738 103254-740 103254-742
Transfer Membranes, Advansta
Transfer Membranes
Nitrocellulose and PVDF membranes for Western blotting.
  • High performance: quality controlled for background-free Western blots
  • Conveniently available as pre-cut or rolls
  • High signal-to-noise ratio for clearer results
  • Flexibility to choose the membrane right for your applications
Advansta offers a large selection of nitrocellulose and polyvinylidene difluoride (PVDF) transfer membranes for immunoblotting applications
The WesternBright PVDF membranes provide durability and high tensile strength for ease of handling. Advansta offers PVDF membranesMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty