Protein Sample Loading Buffers, Advansta
Supplier: Heliszolg kft
R-03018-B10 R-03018-B50 R-03019-B10 R-03019-B50
103254-792EA 21.92 USD InStock
103254-792 103254-794 103254-914 103254-916
Protein Sample Loading Buffers, Advansta
Buffers Electrophoresis Buffers Loading Buffers
Pre-mixed standard Laemmli loading buffers for protein electrophoresis.
  • Pre-mixed - just add equal volume to your protein sample
  • Obtain reproducible results with standardized buffers
  • Choose from reducing or non-reducing loading buffer
These ready-to-use Laemmli loading buffers contain Bromophenol blue to enable visualization of the dye front and monitoring of the progress of electrophoresis.
Choose from reducing or non-reducing buffers, each supplied as a 2X concentrate.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty