Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CRTAM Rabbit Polyclonal Antibody

Supplier: Sino Biological
11975-T16-50 11975-T16-100 11975-T16-200
103587-548EA 142.96 USD InStock
103587-548 103609-010 103609-012
Anti-CRTAM Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human CRTAM (rh CRTAM; Catalog#11975-H08H; NP_062550.2; Met1-Ser286). CRTAM specific IgG was purified by Human CRTAM affinity chromatography.

Type: Primary
Antigen: CRTAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty