Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-ICAM3 Rabbit Monoclonal Antibody [clone: 002]

Supplier: Sino Biological
10333-R002-50 10333-R002-100
103640-286EA 150.18 USD InStock
103640-286 103648-294
Anti-ICAM3 Rabbit Monoclonal Antibody [clone: 002]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD50 / ICAM-3 (rh CD50 / ICAM-3; Catalog#10333-H08H; CAA49369.1; Met1-His485).

Type: Primary
Antigen: ICAM3
Clonality: Monoclonal
Clone: 002
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty