PureCol® Bovine Collagen Type I
Supplier: Robot-Coupe S.N.C.
5074-35ML
103701-210EA 496.92 USD InStock
103701-210
PureCol® Bovine Collagen Type I
Proteins and Peptides
PureCol® EZ Gel is a ready-to-use collagen solution that forms a firm gel by simply warming to 37°C in an incubator. The product consists of purified Type I collagen at a concentration of 0.5%, (~5 mg/ml), DMEM/F-12 medium and a mixture of L-glutamine and dipeptide (L-alanine-L-glutamine) to provide a long-lasting L-glutamine source for cell culture.
PureCol® EZ Gel is designed to improve gel consistency by pre-formulation of the medium and adjustment of the product to a neutral pH. This product avoids the inconsistencies in the preparation of the gel that can arise through variables of reagent addition, pH adjustment and handling conditions.
PureCol® EZ Gel is ideal for providing a firm gel and can also be used in the preparation of a thin layer for culturing cells. PureCol® EZ Gel collagen is provided in a user-friendly packaging for use and storage. This product is sterile and supplied as a ready to use solution.
  • Great for 3D Hydrogels or 2D Coatings
  • Highly purified collagen
  • Over 15 QC tests to ensure consistency and quality
  • Pre-neutralized solution for easy 3D hydrogels
Precaución: This product is for R&D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty