Anti-RNF158 Rabbit Polyclonal Antibody (Cy5®)
Supplier: Bioss
BS-9230R-CY5
10669-618EA 803.92 USD InStock
10669-618
Anti-RNF158 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain this conserved domain are generally involved in the ubiquitination pathway of protein degradation. SH3RF2 (SH3 domain containing ring finger 2), also known as RNF158, is a 729 amino acid protein with one RING-type zinc finger domain and three SH3 domains. Via its RING-type zinc finger domain, SH3RF2 binds an E2 ubiquitin-conjugating enzyme. This suggests that SH3RF2 functions as an E3 ubiquitin-More Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty