CPR Lifeshield™ Plus, Emergency Medical International
Supplier: EMI - Emergency Medical International
474
10756-152KT 34.81 USD InStock
10756-152
CPR Lifeshield™ Plus, Emergency Medical International
First Aid Supplies CPR Products
CPR Lifeshield™ effectively prevents mouth to mouth contact during CPR emergencies.
  • Translucent vinyl also allows monitoring of the patient
  • Each CPR Lifeshield™ is packaged in a polybag with CPR instructions
This lightweight 6³/₄×6³/₄" face conforming mask features a one-way valve and filter that eliminates blowback or body fluids to the rescuer. It is small enough to easily fit in the pocket, holster or trauma kit. Ideal during CPR class it is also compatible with CPR mannequins and models.
Información para pedidos: Includes 1 CPR Lifeshield™, 1More Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty